Thermo Scientific Chemicals

4-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11650.36
500 g, Each
Thermo Scientific Chemicals

4-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A11650.36
500 g, Each
Quantity
Catalog number: A11650.36
also known as A11650-36
Price (USD)
Price: 161.00
Online price: 138.65
Your price:
Quantity
-

Chemical Identifiers

CAS
96-98-0
IUPAC Name
4-methyl-3-nitrobenzoic acid
Molecular Formula
C8H7NO4
InChI Key
BBEWSMNRCUXQRF-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Appearance (Color)
Whtie to pale yellow to cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%
Form
Powder
Melting Point (clear melt)
186.5-192.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text