2,4-Difluorotoluene, 98+%
2,4-Difluorotoluene, 98+%
2,4-Difluorotoluene, 98+%
2,4-Difluorotoluene, 98+%
Thermo Scientific Chemicals

2,4-Difluorotoluene, 98+%

CAS: 452-76-6 | C7H6F2 | 128.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11667.14
also known as A11667-14
Price (USD)
63.65
Online Exclusive
70.90
Save 7.25 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.90
Save 7.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS452-76-6
IUPAC Name2,4-difluoro-1-methylbenzene
Molecular FormulaC7H6F2
InChI KeyMPXDAIBTYWGBSL-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.4470-1.4510 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only