(+/-)-1,3-Butanediol, 99%
(+/-)-1,3-Butanediol, 99%
(+/-)-1,3-Butanediol, 99%
Thermo Scientific Chemicals

(+/-)-1,3-Butanediol, 99%

CAS: 107-88-0 | C4H10O2 | 90.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A11684.0F
also known as A11684-0F
Price (USD)
99.30
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
99.30
Each
Chemical Identifiers
CAS107-88-0
IUPAC Namebutane-1,3-diol
Molecular FormulaC4H10O2
InChI KeyPUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILESCC(O)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4385-1.4415 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Optical Rotation0.0 ± 0.2? (neat)
Appearance (Color)Clear colorless
View more
(±)-1,3-Butanediol acts as a co-monomer in the production of polyurethane and polyester resins. It is used as a humectant (to prevent loss of moisture) in cosmetics, especially in hair sprays and setting lotions. It is used in surfactants, inks, solvents for natural and synthetic flavorings. It is involved in the synthesis of dual peroxisome proliferator-activated gamma and delta agonists acting as euglycemic agents, which is used in the treatment of diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-1,3-Butanediol acts as a co-monomer in the production of polyurethane and polyester resins. It is used as a humectant (to prevent loss of moisture) in cosmetics, especially in hair sprays and setting lotions. It is used in surfactants, inks, solvents for natural and synthetic flavorings. It is involved in the synthesis of dual peroxisome proliferator-activated gamma and delta agonists acting as euglycemic agents, which is used in the treatment of diabetes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, acid chlorides, chloroformates and reducing agents. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Kataoka, N.; Vangnai, A. S.; Pongtharangkul, T.; Tajima, T.; Yakushi, T.; Matsushita, K,; Kato, J. Construction of CoA-dependent 1-butanol synthetic pathway functions under aerobic conditions in Escherichia coli. J. Biotechnol. 2015, 204, 25-32.
  2. Gregory, G. L.; Ulmann, M.; Buchard, A. Synthesis of 6-membered cyclic carbonates from 1,3-diols and low CO2 pressure: a novel mild strategy to replace phosgene reagents. RSC Adv. 2015, 5, 39404-39408.