3-Fluoropyridine, 98%
3-Fluoropyridine, 98%
3-Fluoropyridine, 98%
Thermo Scientific Chemicals

3-Fluoropyridine, 98%

CAS: 372-47-4 | C5H4FN | 97.092 g/mol
Have Questions?
Catalog NumberQuantity
A11693.14
also known as A11693-14
25 g
Catalog number A11693.14
also known as A11693-14
Price (USD)
226.65
Special offer
Online exclusive
Ends: 30-Jun-2026
266.00
Save 39.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
226.65
Special offer
Online exclusive
Ends: 30-Jun-2026
266.00
Save 39.35 (15%)
Each
Chemical Identifiers
CAS372-47-4
IUPAC Name3-fluoropyridine
Molecular FormulaC5H4FN
InChI KeyCELKOWQJPVJKIL-UHFFFAOYSA-N
SMILESFC1=CN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4710-1.4755 @ 20?C
Identification (FTIR)Conforms
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only