2'-Methylacetophenone, 98%
2'-Methylacetophenone, 98%
2'-Methylacetophenone, 98%
Thermo Scientific Chemicals

2'-Methylacetophenone, 98%

CAS: 577-16-2 | C9H10O | 134.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11695.14
also known as A11695-14
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
100.00
Save 14.35 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
100.00
Save 14.35 (14%)
Each
Chemical Identifiers
CAS577-16-2
IUPAC Name1-(2-methylphenyl)ethan-1-one
Molecular FormulaC9H10O
InChI KeyYXWWHNCQZBVZPV-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
Refractive Index1.5295-1.5335 @ 20?C
FormLiquid
2'-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Karl-Heinz Grellmann.; Horst Weller.; Erich Tauer. Tunnel effect on the kinetics of hydrogen shifts. The enol-ketone transformation of 2'-methylacetophenone. Chemical Physics Letters. 1983, 95, (3), 195-199.