4-Ethylbenzoyl chloride, 98+%
4-Ethylbenzoyl chloride, 98+%
4-Ethylbenzoyl chloride, 98+%
4-Ethylbenzoyl chloride, 98+%
Thermo Scientific Chemicals

4-Ethylbenzoyl chloride, 98+%

CAS: 16331-45-6 | C9H9ClO | 168.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Catalog number A11698.09
also known as A11698-09
Price (USD)
45.65
Online Exclusive
50.90
Save 5.25 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.90
Save 5.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS16331-45-6
IUPAC Name4-ethylbenzoyl chloride
Molecular FormulaC9H9ClO
InChI KeyAVTLLLZVYYPGFX-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5460-1.5500 @ 20?C
FormLiquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only