(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water
(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water
(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water
Thermo Scientific Chemicals

(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water

CAS: 2136-75-6 | C20H17OP | 304.329 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number A11709.22
also known as A11709-22
Price (USD)
463.65
Special offer
Online exclusive
Ends: 30-Jun-2026
545.00
Save 81.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
463.65
Special offer
Online exclusive
Ends: 30-Jun-2026
545.00
Save 81.35 (15%)
Each
Chemical Identifiers
CAS2136-75-6
IUPAC Name2-(triphenyl-λ⁵-phosphanylidene)acetaldehyde
Molecular FormulaC20H17OP
InChI KeyCQCAYWAIRTVXIY-UHFFFAOYSA-N
SMILESO=CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Cream to pink or orange to brown
Water Content (Karl Fischer Titration)≤4.0%
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (dry wt. basis)
FormPowder
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

Solubility
Soluble in Chloroform, Methanol.

Notes
Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kenji Kumazawa; Yoshiyuki Wada and Hideki Masuda. Characterization of Epoxydecenal Isomers as Potent Odorants in Black Tea (Dimbula) Infusion.J. Agric. Food Chem. 2006, 54 (13), 4795-4801.
  2. Zh. A. Krasnaya; S. A. Burova; V. S. Bogdanov; Yu. V. Smirnova. Synthesis of δ-hetaryl-α,α-dicarbonylalkadienes and a study of their isomerization.Chemistry of Heterocyclic Compounds. 1997, 33 410-422.
  3. Stable ylide, which undergoes Wittig olefination with aldehydes, but not usually with ketones, to give enals: J. Chem. Soc., 1266 (1961); Angew. Chem. Int. Ed., 14, 486 (1975); Tetrahedron Lett., 3071 (1977). See Appendix 1. Reaction with aldehydes generally gives the thermodynamically more stable (E)-enal. Thus glyoxal gives trans,trans-muconic dialdehyde [(E),(E)-2,4-hexadienal]: Chem. Ber., 107, 710 (1974). The following sequence leads to (Z)-enals: Chem. Ber., 115, 161 (1982):
  4. Wittig reaction followed by reduction with DIBAL-H has been used as a route to allylic alcohols. See, e.g.: J. Org. Chem., 47, 1373 (1982).