5-Methyl-2-nitrophenol, 97%
5-Methyl-2-nitrophenol, 97%
5-Methyl-2-nitrophenol, 97%
Thermo Scientific Chemicals

5-Methyl-2-nitrophenol, 97%

CAS: 700-38-9 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11729.36
also known as A11729-36
Price (USD)
575.65
Online Exclusive
639.00
Save 63.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
575.65
Online Exclusive
639.00
Save 63.35 (10%)
Each
Chemical Identifiers
CAS700-38-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)50.0-60.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only