2,3-Dichloroquinoxaline, 98%
2,3-Dichloroquinoxaline, 98%
2,3-Dichloroquinoxaline, 98%
Thermo Scientific Chemicals

2,3-Dichloroquinoxaline, 98%

CAS: 2213-63-0 | C8H4Cl2N2 | 199.034 g/mol
Have Questions?
Catalog NumberQuantity
A11734.14
also known as A11734-14
25 g
Catalog number A11734.14
also known as A11734-14
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS2213-63-0
IUPAC Name2,3-dichloroquinoxaline
Molecular FormulaC8H4Cl2N2
InChI KeySPSSDDOTEZKOOV-UHFFFAOYSA-N
SMILESClC1=C(Cl)N=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Melting Point (clear melt)149.0-155.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only