Benzyl 4-hydroxybenzoate, 99%
Benzyl 4-hydroxybenzoate, 99%
Benzyl 4-hydroxybenzoate, 99%
Thermo Scientific Chemicals

Benzyl 4-hydroxybenzoate, 99%

CAS: 94-18-8 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A11735.09
also known as A11735-09
Price (USD)
32.65
Online Exclusive
36.20
Save 3.55 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
36.20
Save 3.55 (10%)
Each
Chemical Identifiers
CAS94-18-8
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only