trans-2-Hydroxycinnamic acid, 98+%
trans-2-Hydroxycinnamic acid, 98+%
trans-2-Hydroxycinnamic acid, 98+%
Thermo Scientific Chemicals

trans-2-Hydroxycinnamic acid, 98+%

CAS: 614-60-8 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11740.14
also known as A11740-14
Price (USD)
93.90
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
93.90
Each
Chemical Identifiers
CAS614-60-8
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only