delta-Decanolactone, 97%
delta-Decanolactone, 97%
delta-Decanolactone, 97%
Thermo Scientific Chemicals

delta-Decanolactone, 97%

CAS: 705-86-2 | C10H18O2 | 170.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11756.14
also known as A11756-14
Price (USD)
76.65
Online Exclusive
84.50
Save 7.85 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
84.50
Save 7.85 (9%)
Each
Chemical Identifiers
CAS705-86-2
IUPAC Name6-pentyloxan-2-one
Molecular FormulaC10H18O2
InChI KeyGHBSPIPJMLAMEP-UHFFFAOYNA-N
SMILESCCCCCC1CCCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4555-1.4605 @ 20?C
FormLiquid
Assay (GC)≥96.0%
Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

Solubility
Soluble in water (4 mg/ml at 28°C), alcohols, and propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Korver O. Optical rotatory dispersion and circular dichroism of d-lactones: Determination of the absolute configuration of (+) 5-decanolide and (+) 5-dodecanolide. Tetrahedron. 1970, 26 (10), 2391-96.
  2. Clare J. Veltman; Christian J. Cook; Kelly A. Drake; and Christopher D. Devine . Potential of delta-decanolactone and (Z)-7-dodecen-1-yl acetate to attract feral goats (Capra hircus). Wildlife Research, 2002, 28 (6), 589 - 597.