2-(Methylthio)thiophene, 97%
2-(Methylthio)thiophene, 97%
2-(Methylthio)thiophene, 97%
Thermo Scientific Chemicals

2-(Methylthio)thiophene, 97%

CAS: 5780-36-9 | C5H6S2 | 130.223 g/mol
Have Questions?
Catalog NumberQuantity
A11763.06
also known as A11763-06
5 g
Catalog number A11763.06
also known as A11763-06
Price (USD)
191.65
Online Exclusive
213.00
Save 21.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
191.65
Online Exclusive
213.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS5780-36-9
IUPAC Name2-(methylsulfanyl)thiophene
Molecular FormulaC5H6S2
InChI KeyZLSMPEVZXWDWEK-UHFFFAOYSA-N
SMILESCSC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5925-1.5975 @ 20°C
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhu, D.; Chang, D.; Shi, L. Transition-metal-free cross-coupling of thioethers with aryl(cyano)iodonium triflates: a facile and efficient method for the one-pot synthesis of thiocyanates. Chem. Comm. 2015, 51 (33), 7180-7183.
  2. Yang, C.; Song, H. L.; Chen, F. Response Surface Methodology for Meat-like Odorants from Maillard Reaction with Glutathione I: the Optimization Analysis and the General Pathway Exploration. J. Food Sci. 2012, 77 (9), C966-C974.