Barium sulfate, precipitated, 98%
Barium sulfate, precipitated, 98%
Barium sulfate, precipitated, 98%
Thermo Scientific Chemicals

Barium sulfate, precipitated, 98%

CAS: 7727-43-7 | BaO4S | 233.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A11768.0C
also known as A11768-0C
Price (USD)
346.65
Special offer
Online exclusive
Ends: 30-Jun-2026
408.00
Save 61.35 (15%)
Each
Quantity:
10,000 g
Request bulk or custom format
Price (USD)
346.65
Special offer
Online exclusive
Ends: 30-Jun-2026
408.00
Save 61.35 (15%)
Each
Chemical Identifiers
CAS7727-43-7
IUPAC Namebarium(2+) sulfate
Molecular FormulaBaO4S
InChI KeyTZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES[Ba++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥97.5%
Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, and as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, and as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

Notes
Incompatible with oxidizing agents, aluminum and phosphorus.
RUO – Research Use Only

General References:

  1. Gillani, R.; Ercan, B.; Qiao, A.; Webster, T. J. Nanofunctionalized Zirconia and Barium Sulfate Particles as Bone Cement Additives. Int. J. Nanomedicine 2010, 5, 1-11.
  2. Hennessy, A. J. B.; Graham, G. M. The Effect of Additives on the Co-crystallization of Calcium with Barium Sulphate. J. Cryst. Growth 2002, 237-239, 2153-2159.
  3. Jones, F.; Jones, P.; Ogdena, M. I.; Richmond, W. R.; Rohl, A. L.; Saunders, M. The Interaction of EDTA with Barium Sulfate. J. Colloid. Interf. Sci. 2007, 316, 553-561.