(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%
(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%
(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%
(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%
Thermo Scientific Chemicals

(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%

CAS: 1530-39-8 | C25H21Cl2P | 423.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11769.14
also known as A11769-14
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.70
Save 10.05 (14%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.70
Save 10.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS1530-39-8
IUPAC Name[(4-chlorophenyl)methyl]triphenylphosphanium chloride
Molecular FormulaC25H21Cl2P
InChI KeyRAHOAHBOOHXRDY-UHFFFAOYSA-M
SMILES[Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink
FormPowder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Melting Point (clear melt)282.0-288.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only