Azure II
Azure II
Azure II
Thermo Scientific Chemicals

Azure II

CAS: 37247-10-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A11777.09
also known as A11777-09
Price (USD)
37.65
Online Exclusive
41.20
Save 3.55 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.20
Save 3.55 (9%)
Each
Chemical Identifiers
CAS37247-10-2
SpecificationsSpecification SheetSpecification Sheet
Solution Test(0.1% w/v) : Clear blue solution
Specific Absorbance (E 1%/1cm)(Lambda-max at 244-255nm) : ≥400 (non-U.S. specification)
Specific Absorbance (E 1%/1cm)(Lambda-max at 280-300nm) : ≥800 (non-U.S. specification)
Specific Absorbance (E 1%/1cm)(Lambda-max at 653-665nm) : ≥1500 (non-U.S. specification)
UV spectrophotometry(Lambda-max at 244-255nm) : ≥400 (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only