Sodium metaborate tetrahydrate, 98%
Sodium metaborate tetrahydrate, 98%
Sodium metaborate tetrahydrate, 98%
Sodium metaborate tetrahydrate, 98%
Thermo Scientific Chemicals

Sodium metaborate tetrahydrate, 98%

CAS: 10555-76-7 | BH8NaO6 | 137.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A11803.0C
also known as A11803-0C
Price (USD)
385.65
Special offer
Online exclusive
Ends: 30-Jun-2026
454.00
Save 68.35 (15%)
Each
Quantity:
10,000 g
Request bulk or custom format
Price (USD)
385.65
Special offer
Online exclusive
Ends: 30-Jun-2026
454.00
Save 68.35 (15%)
Each
Chemical Identifiers
CAS10555-76-7
IUPAC Namesodium tetrahydrate oxoborinate
Molecular FormulaBH8NaO6
InChI KeyJAKYJVJWXKRTSJ-UHFFFAOYSA-N
SMILESO.O.O.O.[Na+].[O-]B=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only