Nitromethane, 98+%
Nitromethane, 98+%
Nitromethane, 98+%
Thermo Scientific Chemicals

Nitromethane, 98+%

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11806.22
also known as A11806-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.3800-1.3840 @ 20?C
Assay (GC)≥98.0%
Nitromethane is used as a stabilizer of halogenated organic solvents, rocket and racing fuel and a chemical intermediate. It is also used as a solvent for cyanoacrylate adhesives, polymers and waxes. It serves as a Michael donor, adding to alfa,beta-unsaturated carbonyl compounds through 1,4-addition in the Michael reaction. It acts as a solvent used for extractions, reaction medium and as a cleaning solvent. Further, it is used in the manufacture of pharmaceuticals, explosives, fibers and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitromethane is used as a stabilizer of halogenated organic solvents, rocket and racing fuel and a chemical intermediate. It is also used as a solvent for cyanoacrylate adhesives, polymers and waxes. It serves as a Michael donor, adding to alfa,beta-unsaturated carbonyl compounds through 1,4-addition in the Michael reaction. It acts as a solvent used for extractions, reaction medium and as a cleaning solvent. Further, it is used in the manufacture of pharmaceuticals, explosives, fibers and coatings.

Solubility
Miscible with ethanol, ethyl ether, acetone, carbon tetrachloride and alkali. Slightly miscible with water.

Notes
Incompatible with amines, strong acids, strong bases, strong oxidizing agents, strong reducing agents and copper
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. For conditions for the Henry (nitro-aldol) condensation of nitroalkanes with carbonyl compounds, see 1-Nitropropane, A11975 . In the Tiffeneau-Demjanov ring expansion of ketones, the nitro aldol is reduced to the ß-hydroxyamine before diazotization; see, e.g. (cyclohexanone to cycloheptanone): Org. Synth. Coll., 4, 221 (1963); review: Org. React., 11, 157 (1960).
  2. For a review of the use of nitro compounds as alkyl anion synthons, including methods for removing the nitro group from the resulting carbon skeletons, see: Synthesis, 833 (1988).
  3. For reaction with TosMIC adducts to give pyrroles, see p-Toluenesulfonyl methyl isocyanide, A14312 .
  4. For a brief feature on uses of this reagent in Organic synthesis, see: Synlett, 1174 (2007).
  5. Brequigny, P.; Dayma, G.; Halter, F.; Mounaïm-Rousselle, C.; Dubois, T.; Dagaut, P. Laminar burning velocities of premixed nitromethane/air flames: An experimental and kinetic modeling study. Proc. Combust. Inst. 2015, 35 (1), 703-710.
  6. Wang, C.; Wu, H. L.; Song, Y. F.; He, X.; Yang, Y. Q.; Tan, D. W. Intense pumping and time-and frequency-resolved CARS for driving and tracking structural deformation and recovery of liquid nitromethane molecules. Chem. Phys. Lett. 2015, 640, 101-105.