Lithium L-lactate, 97%
Lithium L-lactate, 97%
Lithium L-lactate, 97%
Thermo Scientific Chemicals

Lithium L-lactate, 97%

CAS: 27848-80-2 | C3H5LiO3 | 96.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A11818.0B
also known as A11818-0B
Price (USD)
549.65
Special offer
Online exclusive
Ends: 30-Jun-2026
647.00
Save 97.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
549.65
Special offer
Online exclusive
Ends: 30-Jun-2026
647.00
Save 97.35 (15%)
Each
Chemical Identifiers
CAS27848-80-2
IUPAC Namelithium(1+) 2-hydroxypropanoate
Molecular FormulaC3H5LiO3
InChI KeyGKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES[Li+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Optical Rotation-14.8? ? 1? (c=1 in water)
Melting Point (clear melt)251-259?C
Appearance (Color)White
View more
Useful chiral synthon, building block for depsipeptides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only