1-Naphthol, 99%
1-Naphthol, 99%
1-Naphthol, 99%
Thermo Scientific Chemicals

1-Naphthol, 99%

CAS: 90-15-3 | C10H8O | 144.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A11862.0E
also known as A11862-0E
Price (USD)
278.65
Special offer
Online exclusive
Ends: 30-Jun-2026
328.00
Save 49.35 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
278.65
Special offer
Online exclusive
Ends: 30-Jun-2026
328.00
Save 49.35 (15%)
Each
Chemical Identifiers
CAS90-15-3
IUPAC Namenaphthalen-1-ol
Molecular FormulaC10H8O
InChI KeyKJCVRFUGPWSIIH-UHFFFAOYSA-N
SMILESOC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White to pale brown or grey
FormCrystals or powder or crystalline powder or flakes
Identification (FTIR)Conforms
Melting point93-98°C
1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthol is used as a precursor in the manufacturing of various azo dyes and pharmaceuticals such as nadolol. It is used as biomarkers. It is used in analytical chemistry as Molisch’s reagent (1-naphthol dissolved in ethanol) for checking the presence of carbohydrates. It plays an essential role with sodium hypobromite to detect the presence of arginine in proteins, which is called as Sakaguchi test.

Solubility
Soluble in benzene, chloroform, ether and ethanol.

Notes
Air and light sensitive. Store in a cool place. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhang, X.; Wang, Y.; Yang, S. Simultaneous removal of Co(II) and 1-naphthol by core-shell structured Fe3O4 @cyclodextrin magnetic nanoparticles. Carbohydr. Polym. 2014, 114, 521-529.
  2. Zuo, L.; Yu, S.; Cheng, L.; Du, E. Adsorption of phenol and 1-naphthol onto XC-72 carbon. Korean J. Chem. Eng. 2013, 30 (3), 714-723.