2-Hydroxy-1,4-naphthoquinone, 98+%
2-Hydroxy-1,4-naphthoquinone, 98+%
2-Hydroxy-1,4-naphthoquinone, 98+%
Thermo Scientific Chemicals

2-Hydroxy-1,4-naphthoquinone, 98+%

CAS: 83-72-7 | C10H6O3 | 174.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A11880.30
also known as A11880-30
Price (USD)
345.65
Online Exclusive
384.00
Save 38.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
345.65
Online Exclusive
384.00
Save 38.35 (10%)
Each
Chemical Identifiers
CAS83-72-7
IUPAC Name4-hydroxy-1,2-dihydronaphthalene-1,2-dione
Molecular FormulaC10H6O3
InChI KeyWVCHIGAIXREVNS-UHFFFAOYSA-N
SMILESOC1=CC(=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to brown
FormPowder
Assay (HPLC)≥98.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Hydroxy-1,4-naphthoquinone is used for preparing decorative hair and skin dyes. 2-Hydroxy-1,4-naphthoquinone also demonstrates antimicrobial and antioxidant effects. It also suppress the formation of hydrogen peroxide and superoxide radical anion by aldehyde oxidase-catalyzed reactions.

Solubility
Soluble in water. 2 g/L at 20°C

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only

General References:

  1. S. Salunke-Gawalia; S.Y. Ranea; V.G. Puranikb; C. Guyard-Duhayonc; F. Varret. Three dimensional hydrogen-bonding network in a copper complex of 2-hydroxy-1,4-naphthoquinone: structural, spectroscopic and magnetic properties. Polyhedron.2004, 23 2541-2547.
  2. Tatsuo Yagishita; Shigeki Sawayama; Ken-ichiro Tsukahara; Tomoko Ogi. Effects of intensity of incident light and concentrations of Synechococcus sp. and 2-hydroxy-1,4-naphthoquinone on the current output of photosynthetic electrochemical cell.Solar Energy.1997,61 347-353.