Michler's Ketone, 98%
Michler's Ketone, 98%
Michler's Ketone, 98%
Thermo Scientific Chemicals

Michler's Ketone, 98%

CAS: 90-94-8 | C17H20N2O | 268.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11889.22
also known as A11889-22
Price (USD)
45.80
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
45.80
Each
Chemical Identifiers
CAS90-94-8
IUPAC Name4-[4-(dimethylamino)benzoyl]-N,N-dimethylaniline
Molecular FormulaC17H20N2O
InChI KeyVVBLNCFGVYUYGU-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)Pale blue to dark blue to dark gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only