1-Iodododecane, 98%, stab. with copper
1-Iodododecane, 98%, stab. with copper
1-Iodododecane, 98%, stab. with copper
Thermo Scientific Chemicals

1-Iodododecane, 98%, stab. with copper

CAS: 4292-19-7 | C12H25I | 296.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11909.14
also known as A11909-14
Price (USD)
42.65
Online Exclusive
46.80
Save 4.15 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
42.65
Online Exclusive
46.80
Save 4.15 (9%)
Each
Chemical Identifiers
CAS4292-19-7
IUPAC Name1-iodododecane
Molecular FormulaC12H25I
InChI KeyGCDPERPXPREHJF-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow to pale orange
Assay (GC)≥97.5%
Refractive Index1.4825-1.4860 @ 20?C
FormLiquid
1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodododecane has been used in the preparation of dodecylated graphite, alkyl functionalized single walled carbon nanotubes (SWNTs). It is also used as chemical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
RUO – Research Use Only

General References:

  1. Soma Chakraborty.; Jayanta Chattopadhyay.; Wenhua Guo.; W Edward Billups. Functionalization of potassium graphite.Angew. Chem. Int. Ed.2007,46(24), 4486-8.
  2. Robin E Anderson.; Andrew R Barron. Solubilization of single-wall carbon nanotubes in Organic solvents without sidewall functionalization.J Nanosci Nanotechnol.2007,7(10), 3436-40.