Methyl vinyl ketone, tech. 90%, stab.
Methyl vinyl ketone, tech. 90%, stab.
Methyl vinyl ketone, tech. 90%, stab.
Thermo Scientific Chemicals

Methyl vinyl ketone, tech. 90%, stab.

CAS: 78-94-4 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11910.AE
also known as A11910-AE
Price (USD)
118.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
118.00
Each
Chemical Identifiers
CAS78-94-4
IUPAC Namebut-3-en-2-one
Molecular FormulaC4H6O
InChI KeyFUSUHKVFWTUUBE-UHFFFAOYSA-N
SMILESCC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer≤0.7% Acetonitrile
Stabilizer0.2-0.5% Hydroquinone
Stabilizer0.2-1.0% Acetic Acid
FormLiquid
Assay (GC)≥80.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only