2-Bromoacetamide, 98%
2-Bromoacetamide, 98%
2-Bromoacetamide, 98%
Thermo Scientific Chemicals

2-Bromoacetamide, 98%

CAS: 683-57-8 | C2H4BrNO | 137.964 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11926.14
also known as A11926-14
Price (USD)
154.65
Online Exclusive
172.00
Save 17.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
154.65
Online Exclusive
172.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS683-57-8
IUPAC Name2-bromoacetamide
Molecular FormulaC2H4BrNO
InChI KeyJUIKUQOUMZUFQT-UHFFFAOYSA-N
SMILESNC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to cream
Assay (GC)≥97.5%
Melting Point (clear melt)85.5-91.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only