Methyl 3,4,5-trimethoxybenzoate, 98+%
Methyl 3,4,5-trimethoxybenzoate, 98+%
Methyl 3,4,5-trimethoxybenzoate, 98+%
Thermo Scientific Chemicals

Methyl 3,4,5-trimethoxybenzoate, 98+%

CAS: 1916-07-0 | C11H14O5 | 226.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11933.36
also known as A11933-36
Price (USD)
98.65
Special offer
Online exclusive
Ends: 30-Jun-2026
116.00
Save 17.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
98.65
Special offer
Online exclusive
Ends: 30-Jun-2026
116.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS1916-07-0
IUPAC Namemethyl 3,4,5-trimethoxybenzoate
Molecular FormulaC11H14O5
InChI KeyKACHFMOHOPLTNX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(OC)=C(OC)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting Point (clear melt)79.0-87.0°C
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only