3-Chlorophenyl isothiocyanate, 97%
3-Chlorophenyl isothiocyanate, 97%
3-Chlorophenyl isothiocyanate, 97%
Thermo Scientific Chemicals

3-Chlorophenyl isothiocyanate, 97%

CAS: 2392-68-9 | C7H4ClNS | 169.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A11999.06
also known as A11999-06
Price (USD)
37.65
Online Exclusive
42.20
Save 4.55 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
42.20
Save 4.55 (11%)
Each
Chemical Identifiers
CAS2392-68-9
IUPAC Name1-chloro-3-isothiocyanatobenzene
Molecular FormulaC7H4ClNS
InChI KeyWGXCKFMVBAOIFH-UHFFFAOYSA-N
SMILESClC1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
FormLiquid
Assay (GC)>96.0%
Refractive Index1.6565-1.6625 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only