2-Chlorothiophenol, 98%
2-Chlorothiophenol, 98%
2-Chlorothiophenol, 98%
Thermo Scientific Chemicals

2-Chlorothiophenol, 98%

CAS: 6320-03-2 | C6H5ClS | 144.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
250 g
Catalog number A12014.06
also known as A12014-06
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.10
Save 5.45 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.10
Save 5.45 (14%)
Each
Chemical Identifiers
CAS6320-03-2
IUPAC Name2-chlorobenzene-1-thiol
Molecular FormulaC6H5ClS
InChI KeyPWOBDMNCYMQTCE-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.6015-1.6055 @ 20°C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only