2-Isopropylphenol, 98+%
2-Isopropylphenol, 98+%
2-Isopropylphenol, 98+%
Thermo Scientific Chemicals

2-Isopropylphenol, 98+%

CAS: 88-69-7 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12027.18
also known as A12027-18
Price (USD)
38.65
Online Exclusive
43.20
Save 4.55 (11%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
43.20
Save 4.55 (11%)
Each
Chemical Identifiers
CAS88-69-7
IUPAC Name2-(propan-2-yl)phenol
Molecular FormulaC9H12O
InChI KeyCRBJBYGJVIBWIY-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange
FormLiquid
Assay (GC)≥98.0%
CommentMay darken on storage
Refractive Index1.5255-1.5295 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only