4-Ethylcatechol, 98%
4-Ethylcatechol, 98%
4-Ethylcatechol, 98%
Thermo Scientific Chemicals

4-Ethylcatechol, 98%

CAS: 1124-39-6 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number A12048.03
also known as A12048-03
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 28-Dec-2026
112.00
Save 16.35
Each
Chemical Identifiers
CAS1124-39-6
IUPAC Name4-ethylbenzene-1,2-diol
Molecular FormulaC8H10O2
InChI KeyHFLGBNBLMBSXEM-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Melting Point (clear melt)36.5-42.5?C
FormFused solid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only