2-Bromo-1,4-dichlorobenzene, 98%
2-Bromo-1,4-dichlorobenzene, 98%
2-Bromo-1,4-dichlorobenzene, 98%
Thermo Scientific Chemicals

2-Bromo-1,4-dichlorobenzene, 98%

CAS: 1435-50-3 | C6H3BrCl2 | 225.894 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12056.06
also known as A12056-06
Price (USD)
40.65
Online Exclusive
44.70
Save 4.05 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS1435-50-3
IUPAC Name2-bromo-1,4-dichlorobenzene
Molecular FormulaC6H3BrCl2
InChI KeyOVXVQBCRONSPDC-UHFFFAOYSA-N
SMILESClC1=CC=C(Cl)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless to cream to pale brown
Melting Point (clear melt)30.0-37.0?C
FormCrystals or powder or crystalline powder or clear liquid as melt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only