2-Amino-5-chloropyridine, 98%
2-Amino-5-chloropyridine, 98%
2-Amino-5-chloropyridine, 98%
Thermo Scientific Chemicals

2-Amino-5-chloropyridine, 98%

CAS: 1072-98-6 | C5H5ClN2 | 128.559 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12063.14
also known as A12063-14
Price (USD)
52.30
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
52.30
Each
Chemical Identifiers
CAS1072-98-6
IUPAC Name5-chloropyridin-2-amine
Molecular FormulaC5H5ClN2
InChI KeyMAXBVGJEFDMHNV-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)133.0-141.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only