3-Acetylpyrrole, 97%
3-Acetylpyrrole, 97%
3-Acetylpyrrole, 97%
Thermo Scientific Chemicals

3-Acetylpyrrole, 97%

CAS: 1072-82-8 | C6H7NO | 109.128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A12073.06
also known as A12073-06
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS1072-82-8
IUPAC Name1-(1H-pyrrol-3-yl)ethan-1-one
Molecular FormulaC6H7NO
InChI KeyKHHSXHXUQVNBGA-UHFFFAOYSA-N
SMILESCC(=O)C1=CNC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (GC)≥96.0%
Melting Point (clear melt)108.0-116.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only