Diethyl methylmalonate, 99%
Diethyl methylmalonate, 99%
Diethyl methylmalonate, 99%
Diethyl methylmalonate, 99%
Thermo Scientific Chemicals

Diethyl methylmalonate, 99%

CAS: 609-08-5 | C8H14O4 | 174.196 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12078.0B
also known as A12078-0B
Price (USD)
260.65
Special offer
Online exclusive
Ends: 30-Jun-2026
307.00
Save 46.35 (15%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
260.65
Special offer
Online exclusive
Ends: 30-Jun-2026
307.00
Save 46.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS609-08-5
IUPAC Name1,3-diethyl 2-methylpropanedioate
Molecular FormulaC8H14O4
InChI KeyUPQZOUHVTJNGFK-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to very pale yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4115-1.4145 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only