1-Iodo-2-nitrobenzene, 97%
1-Iodo-2-nitrobenzene, 97%
1-Iodo-2-nitrobenzene, 97%
Thermo Scientific Chemicals

1-Iodo-2-nitrobenzene, 97%

CAS: 609-73-4 | C6H4INO2 | 249.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12100.14
also known as A12100-14
Price (USD)
63.65
Online Exclusive
70.70
Save 7.05 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.70
Save 7.05 (10%)
Each
Chemical Identifiers
CAS609-73-4
IUPAC Name1-iodo-2-nitrobenzene
Molecular FormulaC6H4INO2
InChI KeyJXMZUNPWVXQADG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥96.0%
FormPowder mixed with darker coarse granules and/or lumps
Melting Point (clear melt)48-53?C
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only