1-Bromo-4-(trifluoromethoxy)benzene, 98%
1-Bromo-4-(trifluoromethoxy)benzene, 98%
1-Bromo-4-(trifluoromethoxy)benzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-(trifluoromethoxy)benzene, 98%

CAS: 407-14-7 | C7H4BrF3O | 241.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12104.14
also known as A12104-14
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS407-14-7
IUPAC Name1-bromo-4-(trifluoromethoxy)benzene
Molecular FormulaC7H4BrF3O
InChI KeySEAOBYFQWJFORM-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4595-1.4635 @ 20?C
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only