2,6-Dinitroaniline, 98%
2,6-Dinitroaniline, 98%
2,6-Dinitroaniline, 98%
Thermo Scientific Chemicals

2,6-Dinitroaniline, 98%

CAS: 606-22-4 | C6H5N3O4 | 183.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12120.14
also known as A12120-14
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS606-22-4
IUPAC Name2,6-dinitroaniline
Molecular FormulaC6H5N3O4
InChI KeyQFUSCYRJMXLNRB-UHFFFAOYSA-N
SMILESNC1=C(C=CC=C1[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to yellow-brown to brown
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)136.0-142.0?C
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dinitroaniline is used for organic synthesis intermediates, dyes intermediates, used in the preparation of 2,6-dinitroiodobenzene. It act as a precursor for various new 1,2,3-trisubstituted halobenzene derivatives.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from strong acids, acid chlorides, acid anhydrides.
RUO – Research Use Only