Thermo Scientific Chemicals

AmberChrom 1x2 50-100 (Cl), Thermo Scientific Chemicals

Catalog number: A12130.30
250 g, Each
Thermo Scientific Chemicals

AmberChrom 1x2 50-100 (Cl), Thermo Scientific Chemicals

Catalog number: A12130.30
250 g, Each
Quantity
Catalog number: A12130.30
also known as A12130-30
Price (USD)
Price: 162.00
Online price: 137.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9085-42-1
MDL Number
MFCD00132715
Appearance (Color)
White to pale brown
Form
Beads
Comment
Ionic form: Cl⁻
Comment
Total exchange capacity: 0.7 meq/ml
Particle Size
50-100 mesh (dry)

Description

It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text