4-tert-Butylthiophenol, 97%
4-tert-Butylthiophenol, 97%
4-tert-Butylthiophenol, 97%
4-tert-Butylthiophenol, 97%
Thermo Scientific Chemicals

4-tert-Butylthiophenol, 97%

CAS: 2396-68-1 | C10H14S | 166.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12148.14
also known as A12148-14
Price (USD)
317.65
Online Exclusive
353.00
Save 35.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
317.65
Online Exclusive
353.00
Save 35.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2396-68-1
IUPAC Name4-tert-butylbenzene-1-thiol
Molecular FormulaC10H14S
InChI KeyGNXBFFHXJDZGEK-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5455-1.5505 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
4-tert-butylthiophenol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-butylthiophenol is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Feng Wang & Masahiro Tada. Asymmetric Michael Addition with Amino Alcohol Catalysts Derived from d-Glucose. Agricultural and Biological Chemistry. 1990, 54 (11), 370-375.
  2. Paul McDaid; Yonggang Chen and Li Deng Dr. A Highly Enantioselective and General Conjugate Addition of Thiols to Cyclic Enones with an Organic Catalyst. ACS Catalysis. 2002, 114 (2), 348-350.