1-Bromo-4-chlorobenzene, 98+%
1-Bromo-4-chlorobenzene, 98+%
1-Bromo-4-chlorobenzene, 98+%
Thermo Scientific Chemicals

1-Bromo-4-chlorobenzene, 98+%

CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1000 g
Catalog number A12160.22
also known as A12160-22
Price (USD)
27.65
Online Exclusive
30.60
Save 2.95 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.60
Save 2.95 (10%)
Each
Chemical Identifiers
CAS106-39-8
IUPAC Name1-bromo-4-chlorobenzene
Molecular FormulaC6H4BrCl
InChI KeyNHDODQWIKUYWMW-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)63.0-69.0?C
FormCrystals or powder or crystalline powder or chunks or fused solid
Appearance (Color)White to pale cream to black
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 0.045g/L.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Harry A.J. Oonka; Peter R. van der Lindeb; Jan Huinink; Jacobus G. Blok. Representation and assessment of vapor pressure data; a novel approach applied to crystalline 1-bromo-4-chlorobenzene, 1-chloro-4-iodobenzene, and 1-bromo-4-iodobenzene. The Journal of Chemical Thermodynamics. 1998, 30 (7), 897-907.
  2. Vincenzo Calò; Angelo Nacci; Antonio Monopoli Dr.; and Pietro Cotugno. Heck Reactions with Palladium Nanoparticles in Ionic Liquids: Coupling of Aryl Chlorides with Deactivated Olefins. Angewandte Chemie. 2009, 121 (33), 6217-6219.