2-Chloro-3-nitropyridine, 99%
2-Chloro-3-nitropyridine, 99%
2-Chloro-3-nitropyridine, 99%
Thermo Scientific Chemicals

2-Chloro-3-nitropyridine, 99%

CAS: 5470-18-8 | C5H3ClN2O2 | 158.541 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12161.06
also known as A12161-06
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.30
Save 5.65 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.30
Save 5.65 (15%)
Each
Chemical Identifiers
CAS5470-18-8
IUPAC Name2-chloro-3-nitropyridine
Molecular FormulaC5H3ClN2O2
InChI KeyUUOLETYDNTVQDY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)98.0-105.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only