N-Methylethylenediamine, 95%
N-Methylethylenediamine, 95%
N-Methylethylenediamine, 95%
N-Methylethylenediamine, 95%
Thermo Scientific Chemicals

N-Methylethylenediamine, 95%

CAS: 109-81-9 | C3H12N2 | 76.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12165.14
also known as A12165-14
Price (USD)
219.65
Online Exclusive
244.00
Save 24.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
219.65
Online Exclusive
244.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS109-81-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4375-1.4435 @ 20?C
Assay (GC)≥94.0%
Identification (FTIR)Conforms
N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Sandip Mukherjee and Partha Sarathi Mukherjee. Cu(II)-azide polynuclear complexes of Cu4 building clusters with Schiff-base co-ligands: synthesis, structures, magnetic behavior and DFT studies. Dalton Transactions. 2013, 42 (11), 4019-4030.
  2. Neil Vasdev; Frank J. LaRonde; James R. Woodgett; Armando Garcia; Elizabeth A. Rubie; Jeffrey H. Meyer; Sylvain Houle; Alan A. Wilson; Neil Vasdev; Frank J. LaRonde; James R. Woodgett; Armando Garcia; Elizabeth A. Rubie; Jeffrey H. Meyer; Sylvain Houle; Alan A. Wilson; Neil Vasdev; Frank J. LaRonde; James R. Woodgett; Armando Garcia; Elizabeth A. Rubie, Jeffrey H. Meyer; Sylvain Houle; Alan A.Wilson. Rationally designed PKA inhibitors for positron emission tomography: Synthesis and cerebral biodistribution ofN-(2-(4-bromocinnamylamino)ethyl)-N-[11C]methyl-isoquinoline-5-sulfonamide. Bioorganic & Medicinal Chemistry. 2008, 16, (9), 5277-5284.