Sodium hydrogen selenite, 96%
Sodium hydrogen selenite, 96%
Sodium hydrogen selenite, 96%
Sodium hydrogen selenite, 96%
Thermo Scientific Chemicals

Sodium hydrogen selenite, 96%

CAS: 7782-82-3 | Na2O3Se | 172.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12173.18
Price (USD)
51.65
Special offer
Online exclusive
Ends: 31-Dec-2025
60.60
Save 8.95 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 31-Dec-2025
60.60
Save 8.95 (15%)
Each
Add to cart
Chemical Identifiers
CAS7782-82-3
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis(Selenium, Se) : 49.69-54.91% (Theory 52.31%)
Assay (unspecified)(By Selenium Elemental Analysis) : >95.0 to <105.0%
Appearance (Color)White to pale cream to pale gray
FormPowder
Sodium hydrogen selenite is used as a culture media additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen selenite is used as a culture media additive.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. F. Badii; M. O. Moss and K. Wilson. The effect of sodium biselenite on the growth and aflatoxin production of Aspergillus parasiticus and the growth of other aspergilli. Letters in Applied Microbiology. 1986, 2 (4), 61-64.
  2. Johnson, L.P.V. Embryonic reaction to sodium biselenite as a test of seed vitality. Angewandte Chemie. 2009, 39 (11), 943-947.