2',5'-Dihydroxyacetophenone, 98+%
2',5'-Dihydroxyacetophenone, 98+%
2',5'-Dihydroxyacetophenone, 98+%
Thermo Scientific Chemicals

2',5'-Dihydroxyacetophenone, 98+%

CAS: 490-78-8 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12185.22
also known as A12185-22
Price (USD)
331.65
Online Exclusive
368.00
Save 36.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
331.65
Online Exclusive
368.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS490-78-8
IUPAC Name1-(2,5-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O3
InChI KeyWLDWSGZHNBANIO-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥98.0%
Melting Point (clear melt)203.5-209.5?C
Appearance (Color)Yellow to green
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only