5-Bromo-2-chlorobenzoic acid, 98+%
5-Bromo-2-chlorobenzoic acid, 98+%
5-Bromo-2-chlorobenzoic acid, 98+%
5-Bromo-2-chlorobenzoic acid, 98+%
Thermo Scientific Chemicals

5-Bromo-2-chlorobenzoic acid, 98+%

CAS: 21739-92-4 | C7H4BrClO2 | 235.461 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12218.30
also known as A12218-30
Price (USD)
313.65
Special offer
Online exclusive
Ends: 30-Jun-2026
369.00
Save 55.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
313.65
Special offer
Online exclusive
Ends: 30-Jun-2026
369.00
Save 55.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS21739-92-4
IUPAC Name5-bromo-2-chlorobenzoate
Molecular FormulaC7H3BrClO2
InChI KeyFGERXQWKKIVFQG-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC(Br)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)155.0-162.0?C
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Appearance (Color)White
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

Solubility
Soluble in water 2.63 g/L @20°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Michael Tran; Qing Sun; Benjamin W. Smitha and James D. Winefordner. Determination of C: H: O: N ratios in solid organic compounds by laser-induced plasma spectroscopy. J. Anal. At. Spectrom. 2001, 16 (6), 628-632.
  2. Michael Tran; Qing Sun; Benjamin W. Smith and James D. Determination of F, Cl, and Br in Solid Organic Compounds by Laser-Induced Plasma Spectroscopy. Applied Spectroscopy. 2001, 55 (6), 739-744.