Tyramine hydrochloride, 98%
Tyramine hydrochloride, 98%
Tyramine hydrochloride, 98%
Tyramine hydrochloride, 98%
Thermo Scientific Chemicals

Tyramine hydrochloride, 98%

CAS: 60-19-5 | C8H12ClNO | 173.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12220.14
also known as A12220-14
Price (USD)
87.65
Online Exclusive
96.80
Save 9.15 (9%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
96.80
Save 9.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS60-19-5
IUPAC Namehydrogen 4-(2-aminoethyl)phenol chloride
Molecular FormulaC8H12ClNO
InChI KeyRNISDHSYKZAWOK-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)271.0-277.0?C
Reagent for fluorescence enzyme immunoassay

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only