1-Hexyl isothiocyanate, 97%
1-Hexyl isothiocyanate, 97%
1-Hexyl isothiocyanate, 97%
Thermo Scientific Chemicals

1-Hexyl isothiocyanate, 97%

CAS: 4404-45-9 | C7H13NS | 143.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12223.06
also known as A12223-06
Price (USD)
59.30
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
59.30
Each
Chemical Identifiers
CAS4404-45-9
IUPAC Name1-isothiocyanatohexane
Molecular FormulaC7H13NS
InChI KeyWXYAXKKXIGHXDS-UHFFFAOYSA-N
SMILESCCCCCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4900-1.4950 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only