Rubidium carbonate, 99%
Rubidium carbonate, 99%
Rubidium carbonate, 99%
Thermo Scientific Chemicals

Rubidium carbonate, 99%

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12240.06
also known as A12240-06
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.80
Save 6.15 (16%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 30-Jun-2026
38.80
Save 6.15 (16%)
Each
Chemical Identifiers
CAS584-09-8
IUPAC Namedirubidium(1+) carbonate
Molecular FormulaCO3Rb2
InChI KeyWPFGFHJALYCVMO-UHFFFAOYSA-L
SMILES[Rb+].[Rb+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)> 98.5 to <101.5%
FormPowder
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.

Solubility
Soluble in water4500 g/L @20°C.

Notes
Hygroscopic. Store under dry inert gas. Store away from bases, water/ moisture.
RUO – Research Use Only

General References:

  1. Kenji Toda and Mineo Sato. Synthesis and structure determination of new layered perovskite compounds, ALaTa2O7 and ACa2Ta3O10(A = Rb, Li). J. Mater. Chem. 2007, 6 (6), 1067-1071.
  2. Toshihiko Honma; Kenji Toda; Zuo-Guang Ye; Mineo Sato. Concentration quenching of the eu3+-activated luminescence in some layered perovskites with two-dimensional arrangement. Talanta. 1998, 59 (8), 1187-1193.