N-(Hydroxymethyl)benzamide, 98%
N-(Hydroxymethyl)benzamide, 98%
N-(Hydroxymethyl)benzamide, 98%
N-(Hydroxymethyl)benzamide, 98%
Thermo Scientific Chemicals

N-(Hydroxymethyl)benzamide, 98%

CAS: 6282-02-6 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
250 g
Catalog number A12250.30
also known as A12250-30
Price (USD)
415.65
Special offer
Online exclusive
Ends: 30-Jun-2026
489.00
Save 73.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
415.65
Special offer
Online exclusive
Ends: 30-Jun-2026
489.00
Save 73.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS6282-02-6
IUPAC NameN-(hydroxymethyl)benzamide
Molecular FormulaC8H9NO2
InChI KeyUOUBPDZUBVJZOQ-UHFFFAOYSA-N
SMILESOCNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Melting Point (clear melt)97.0-104.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only