1-Bromo-3-ethylbenzene, 98%
1-Bromo-3-ethylbenzene, 98%
1-Bromo-3-ethylbenzene, 98%
1-Bromo-3-ethylbenzene, 98%
Thermo Scientific Chemicals

1-Bromo-3-ethylbenzene, 98%

CAS: 2725-82-8 | C8H9Br | 185.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12258.03
also known as A12258-03
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.60
Save 3.95 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.60
Save 3.95 (14%)
Each
Add to cart
Chemical Identifiers
CAS2725-82-8
IUPAC Name1-bromo-3-ethylbenzene
Molecular FormulaC8H9Br
InChI KeyZRFJYAZQMFCUIX-UHFFFAOYSA-N
SMILESCCC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.5430-1.5470 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only